Learn More
CERILLIANT™ Guaifenesin Impurity Mixture Solution, 0.5-1.0 mg/mL in Acetonitrile (each analyte), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco G0221ML
Catalog No. 01-001-407
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Guaifenesin, a glyceryl ether of guaiacol, is a key constituent in cold and cough formulations. It belongs to the group of expectorant medicines because of its ability to ease the discomfort caused due to cough by causing an increase in the sputum volume and a simultaneous decrease in its viscosity. Its applications include short term relief in the breathing passageway in various conditions such as allergy, cold, and upper respiratory tract infections. It has also been proposed to be effective in the management of pain in various conditions such as prostatitis, seminal vesiculitis, benign prostatic hypertrophy, prostatic abscess, bladder neck hypertrophy, dysmenorrhea, and urinary tract infection.
Specifications
Specifications
75-05-8
UN 1648 3 / PGII
124.14
-20°C
2°C (35.6°F) - closed cup
C7H8O2
0.5 to 1.0 mg/mL in acetonitrile (each analyte)
Ampule
1 mL
Safety and Handling
Safety and Handling
H332-H319
H312
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation