Learn More
CERILLIANT™ Iloperidone-13C,D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco I0131ML
Catalog No. 01-001-487
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This internal standard is suitable for Iloperidone testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, forensic analysis, or urine drug testing. Iloperidone is an atypical antipsychotic administered for the treatment of schizophrenia. The drug is marketed under the trade names Fanapt and Fanapta.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
430.49
-20°C
9.7°C (49.5°F)
C2313CH24D3FN2O4
100 μg/mL in methanol
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation