Learn More
CERILLIANT™ Indeno[1,2,3-cd]pyrene, , Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco SCI0011G
Description
Indeno[1,2,3-cd]pyrene belongs to the class of polycyclic aromatic hydrocarbons (PAHs), also called as polynuclear aromatic hydrocarbons or polyarenes, which are regarded as potential pollutants in the environment. PAHs are typically produced due to natural as well as anthropogenic activities; incomplete combustion of the organic matter being the predominant cause.Specifications
193-39-5 | |
MFCD00152577 | |
Vial | |
1 g |
C22H12 | |
276.33 | |
Room temp |
Safety and Handling
EINECSNumber : 205-893-2
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation