Learn More
CERILLIANT™ Isobutyryl fentanyl-13C6 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco I0410.5ML
Description
Isobutyryl fentanyl is a fentanyl analog and sold on the illicit market as a designer drug or in combination with heroin. This stable-labeled internal standard is suitable for quantification of isobutyryl fentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.Specifications
67-56-1 | |
100 μg/mL in methanol (as free base) | |
Ampule | |
0.5 mL |
C1713C6H30N2O · HCl | |
392.91 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation