Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ MAM2201, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco S0911ML
Description
A certified solution standard suitable for use in LC/MS or GC/MS synthetic cannabinoid testing applications including clinical toxicology, forensic analysis, and urine drug testing. MAM2201 is a hybrid of the synthetic cannabinoids JWH-122 and AM-2201. This drug has been identified in European synthetic cannabinoid blends.Specifications
67-56-1, 1354631-24-5 | |
MFCD22421647 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C25H24FNO | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
373.46 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation