Learn More
CERILLIANT™ Mazindol Solution, 1 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco M2071ML
Catalog No. 01-001-679
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Mazindol, sold as Mazanor or Sanorex, is a sympathomimetic amine stimulant drug similar to amphetamine which is used as an appetite suppressant. Mazindol is not currently available as a commercially marketed and FDA regulated prescription agent for the treatment of obesity. This Snap-N-Spike™ Reference Solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as forensic analysis, urine drug testing and clinical toxicology testing.
Specifications
Specifications
67-56-1, 22232-71-9
UN 1230 - class 3 - PG 2 - Methanol, solution
284.74
-20°C
9.7°C (49.5°F) - closed cup
C16H13ClN2O
1 mg/mL in methanol
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation