Learn More
CERILLIANT™ MDPPP Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco M1761ML
Catalog No. 01-001-656
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
A Certified Spiking Solution™ suitable for use as starting material in calibrators and controls for LC/MS or GC/MS MDPPP testing methods in clinical toxicology, forensic analysis, or urine drug testing applications. 3’,4’-Methylenedioxy-a-pyrrolidinopropiophenone, also known as MDPPP, is a synthetic cathinone found in numerous bath salt blends under the street name NRG-1. This designer drug first emerged in Germany in the late 1990s and early 2000s as an ingredient in imitation ecstasy pills.
Specifications
Specifications
67-56-1, 24698-57-5
UN 1230 - class 3 - PG 2 - Methanol
3′,4′-Methylenedioxy-alpha-pyrrolidinopropiophenone hydrochloride solution
Ampule
1 mL
C14H17NO3 · HCl
1.0 mg/mL in methanol (as free base)
283.75
-20°C
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation