Learn More
CERILLIANT™ Meclonazepam-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1981ML
Description
An internal standard suitable for use in LC/MS or GC/MS testing methods of meclonazepam for clinical toxicology, forensic analysis, urine drug testing or isotope dilution methods. Meclonazepam is a benzodiazepine derivative similar in structure to clonazepam, but was never used as medicine and instead appeared on the illicit drug market as a designer drug.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
332.76 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C16H9D3ClN3O3 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation