Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Mescaline Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco M0471ML

Catalog No. 01-001-589

Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

A certified solution standard applicable for use in a variety of LC/MS or GC/MS applications from urine drug testing to forensic analysis and clinical toxicology. Mescaline is an illicit psychedelic drug also known as Peyote. This alkaloid occurs naturally in the peyote cactus.
Specifications

Specifications

67-56-1, 832-92-8
UN 1230 - class 3 - PG 2 - Methanol, solution
247.72
1 mL
C11H17NO3·HCl
1.0 mg/mL in methanol (as free base)
Ampule
9.7°C (49.5°F) - closed cup
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

CERILLIANT is a registered trademark of Cerilliant Corporation