Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Metaxalone-D6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1431ML
Description
Metaxalone, also known as Skelaxin™, is a skeletal muscle relaxant that also relieves pain caused by strains, sprains, and other musculoskeletal conditions. This new internal standard is suitable for numerous LC/MS or GC/MS applications from forensic testing, clinical toxicology, or pain prescription monitoring to urine drug testing and isotope dilution methods.Specifications
67-56-1 | |
MFCD08064199 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C12H9NO3D6 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
227.29 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation