Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ (+/-)-Methadone Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M0191ML
Description
Methadone is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. The drug is sold under several trade names such as Symoron, Dolophine™, Amidone, Methadose™, Physeptone, or Heptadon. This Certified Spiking Solution™ is suitable for LC/MS or GC/MS testing methods in pain prescription monitoring, forensic analysis or clinical toxicology.Specifications
67-56-1, 76-99-3 | |
MFCD00056289 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C21H27NO | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
309.45 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation