Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Methohexital-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M0791ML
Description
Methohexital is a short acting barbiturate anesthetic used to induce sedation and anesthesia for medical procedures. This internal standard is suitable for quantitation of methohexital levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis.Specifications
67-56-1, 160227-45-2 | |
MFCD00673295 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C14D5H13N2O3 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
267.34 | |
2 to 8°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation