Learn More
CERILLIANT™ Methoxetamine-D3 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1871ML
Description
Methoxetamine, with street names such as MXE, Mexxy, and Roflcoptr, is a designer drug and ketamine derivative used recreationally as a dissociative and hallucinogen. Methoxetamine use is reportedly popular in the United Kingdom with cases also observed in the US. This stable-labeled internal standard is suitable for use in GC/MS or LC/MS methoxetamine testing methods for applications in clinical toxicology, forensic analysis, or urine drug testing.Specifications
67-56-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
286.81 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C15H18D3NO2 · HCl | |
100 μg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation