Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Mianserin Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M9191ML
Description
Mianserin is a tetracyclic antidepressant prescribed for the treatment of depression and is sold under the trade names Lumin and Tolvon. This certified solution standard is applicable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis. This tetracyclic antidepressant functions as a noradrenergic and a specific serotonergic antidepressant with anxiolytic, hypnotic, antiemetic, orexigenic, and antihistamine effects.Specifications
67-56-1, 21535-47-7 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
300.83 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C18H21ClN2 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation