Learn More
CERILLIANT™ MMB-2201 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  MilliporeSigma Supelco S1291ML
Catalog No. 01-002-019
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
MMB-2201, also known as 5-fluoro AMB-PICA and 5-fluoro MMB-PICA, is a synthetic cannabinoid found in herbal blends. This certified reference solution is suited for use in synthetic cannabinoid testing methods by LC/MS o GC/MS for applications in clinical toxicology, urine drug testing, and forensic analysis.
Specifications
Specifications
75-05-8, 1971007-87-0
100 μg/mL in acetonitrile
Ampule
6°C (42.8°F) - closed cup
C20H27FN2O3
362.44
1 mL
Safety and Handling
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation