Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Mycophenolic acid-beta-D-glucuronide Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M1351ML
Description
A Certified Spiking Solution™ for use in therapeutic drug monitoring analyses of serum or whole blood by HPLC or LCMS. The glucuronide, also known as MPAG, is the primary metabolite of the immunosuppressant mycophenolic acid and is monitored by clinical labs to ensure patients remain within the drug's therapeutic range.Specifications
75-05-8, 31528-44-6 | |
MFCD09878308 | |
1.0 mg/mL in acetonitrile | |
Ampule | |
1 mL |
C23H28O12 | |
UN 1648 3 / PGII | |
496.46 | |
-20°C | |
2°C (35.6°F) - closed cup |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation