Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ N-Desmethylmirtazapine-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco D1641ML
Description
A stable-labeled internal standard suitable for quantitation of N-Desmethylmirtazapine levels in urine, serum, or plasma by LC/MS or GC/MS for forensic analysis, clinical toxicology, or urine drug testing. N-Desmethylmirtazapine is the major urinary metabolite of the tetracyclic antidepressant mirtazapine, marketed under the trade names Remeron™, Avanza, or Zispin for treatment of depression. The metabolite, also known as normirtazapine, is pharmacologically active.Specifications
75-05-8, 1188331-80-7 | |
UN 1648 3 / PGII | |
255.35 | |
1 mL |
C16H13D4N3 | |
100 μg/mL in acetonitrile | |
Ampule | |
2°C (35.6°F) - closed cup |
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation