Learn More
CERILLIANT™ N-Desmethyltrimipramine-D3 Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco D1141ML
Catalog No. 01-001-226
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
N-Desmethyltrimipramine is a primary blood metabolite of trimipramine. Trimipramine is a tricyclic antidepressant prescribed for treatment of depression, insomnia, and chronic pain or as an adjunctive therapy for alcohol or opioid withdrawal. This stable-labeled internal standard is suitable for quantification of N-Desmethyltrimipramine levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
319.89
-20°C
9.7°C (49.5°F) - closed cup
C19H21D3N2 · HCl
100 μg/mL in methanol (as free base)
Ampule
1 mL
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation