Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ (+/-)-N-Ethylamphetamine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco E0181ML
Description
N-Ethylamphetamine is a stimulant drug of the phenethylamine and amphetamine classes. This certified solution standard is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from sports testing and clinical toxicology to forensic analysis and urine drug testing. N-Ethylamphetamine, sold as the pharmaceutical appetite-suppressant Apetinil, is also abused as a recreational drug with a stimulant effect similar to that of its amphetamine and methamphetamine analogs.Specifications
67-56-1, 457-87-4 | |
MFCD00801115 | |
1.0 mg/mL in methanol | |
Ampule | |
1 mL |
C11H17N | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
163.26 | |
2 to 8°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation