Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ N-Ethylcathinone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco E0801ML
Description
N-Ethylcathinone, or ethylpropion, is an amphetamine and stimulant drug sold along with mephedrone as 'ecstasy' at parties and raves. With street names such as ETH-CAT, the illicit drug is also sold in bath salt blends across the US, Europe, and Australia. This Certified Spiking Solution™ is suitable as starting material in calibrators and controls for GC/MS or LC/MS bath salt testing applications in forensic analysis, urine drug testing or clinical toxicology.Specifications
67-56-1, 51553-17-4 | |
MFCD00012214 | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C11H15NO · HCl | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
213.70 | |
-20°C | |
9.7°C (49.5°F) |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation