Learn More
CERILLIANT™ N-Ethylhexedrone Hydrochloride Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco E1331ML
Description
N-Ethylhexedrone, also known as a-ethylaminocaprophenone, N-ethylnorhexedrone and NEH, is a stimulant of the cathinone class. A Snap-N-Spike™ reference solution applicable for use in urine drug testing, clinical toxicology, and forensic analysis by LC-MS/MS or GC/MS.Specifications
67-56-1 | |
MFCD30718314 | |
255.78 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C14H21NO · HCl | |
1 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation