Learn More
CERILLIANT™ Nalbuphine-3-beta-D-glucuronide Solution, 100 μg/mL (Methanol:Water (90:10) (v/v)), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco N1321ML
Description
A Snap-N-Spike™ reference solution applicable for use in urine drug testing, pain prescription monitoring, clinical toxicology, forensic analysis, or pharmaceutical research by LC-MS/MS or GC/MS. Nalbuphine glucuronide is the major metabolite of nalbuphine, a semisynthetic opioid analgesic sold under the trade name Nubain™.Specifications
67-56-1, 1634639-65-8 | |
100 μg/mL (methanol:water (90:10) (v/v)) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C27H35NO10 | |
533.57 | |
1 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation