Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Nalorphine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco N9241ML
Description
A certified solution standard applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as clinical toxicology or forensic analysis. Nalorphine is an opiate used to counteract the effects of opioid overdose.Specifications
67-56-1, 57-29-4 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
347.84 | |
1 mL |
C19H21NO3·HCl | |
1.0 mg/mL in methanol (as free base) | |
Ampule | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation