Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Noratropine Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N0571ML
Description
The primary metabolite of atropine, an anticholinergic drug used in opthalmology, resuscitation from cardiac arrest, and treatment for organophosphate poisoning. A Certified Solution Standard for use in LC or GC applications.Specifications
67-56-1, 537-29-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
Noratropine hydrochloride | |
Ampule | |
1 mL |
C16H21NO3·HCl | |
1.0 mg/mL in methanol (as free base) | |
311.80 | |
-20°C | |
9°C (48.2°F) |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation