Learn More
CERILLIANT™ Nordothiepin HCl (cis/trans) Solution, 1 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N1221ML
Description
Nordothiepin, also known as N-desmethyldosulepin and Northiaden, is the major active metabolite of the tricyclic antidepressant dothiepin. Due to high toxicity and risk of overdose dothiepin and its metabolite nordothiepin are assayed in therapeutic drug monitoring to ensure patients remain within the drug's therapeutic range. This certified Snap-N-Spike™ solution is suitable for LC-MS/MS applications including therapeutic drug monitoring, clinical toxicology, urine drug testing or forensic analysis.Specifications
67-56-1, 844-12-2 | |
1 mg/mL in methanol (as free base) | |
Ampule | |
1 mL |
C18H19NS · HCl | |
317.88 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation