Learn More
CERILLIANT™ Nortetrazepam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco N1251ML
Description
Nortetrazepam is a major metabolite of tetrazepam. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for nortetrazepam LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications.Specifications
67-56-1, 10379-11-0 | |
MFCD00868023 | |
274.75 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C15H15ClN2O | |
1.0 mg/mL in methanol | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation