Learn More
CERILLIANT™ o-Fluorofentanyl-13C6 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0770.5ML
Description
This stable-labeled internal standard is suitable for quantification of ortho-Fluorofentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. ortho-Fluorofentanyl, also known as 2-Fluorofentanyl, is an opioid analgesic being an analogue of fentanyl that is sold as designer drug on the illicit drug market.Specifications
67-56-1 | |
100 μg/mL in methanol (as free base) | |
Ampule | |
0.5 mL |
C1613C6H27FN2O · HCl | |
396.88 | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation