Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Omeprazole Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco O0211ML
Description
Omeprazole, sold under the trade name Prilosec™, is a proton pump inhibitor used in the treatment of peptic ulcers and GERD. This Snap-N-Spike™ reference solution is applicable for use in clinical toxicology or forensic analysis by LC-MS/MS or GC/MS.Specifications
67-56-1, 73590-58-6 | |
MFCD00083192 | |
1.0 mg/mL in methanol | |
345.42 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C17H19N3O3S | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
Omeprazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Antra, Losec | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation