Learn More
CERILLIANT™ OPC-3373-D4 (Aripiprazole Metabolite-D4) Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco O0461ML
Catalog No. 01-001-817
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
OPC-3373 is a metabolite of aripiprazole, marketed as the atypical antipsychotic Abilify™ for the treatment of schizophrenia, bipolar disorder, and clinical depression. This stable-labeled internal standard is suitable for quantification of OPC-3373 levels by LC/MS or GC/MS for applications in clinical diagnostic, urine drug testing or forensic analysis.
Specifications
Specifications
75-05-8
UN 1648 3 / PGII
253.29
-20°C
2°C (35.6°F) - closed cup
C13H11D4NO4
100 μg/mL (acetonitrile:DMSO (9:1))
Ampule
1 mL
Safety and Handling
Safety and Handling
H332-H319
H312
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation