Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Oxazepam glucuronide Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco O0231ML
Description
Oxazepam glucuronide is a major urinary metabolite of the benzodiazepine drug, oxazepam. Oxazepam, marketed as Alepam, Medopam, or Murelax™, has been used extensively since the 1960s for treatment of anxiety and insomnia and in the control of symptoms of alcohol withdrawal. This certified reference solution is suitable for GC/MS or LC/MS applications with oxazepam or oxazepam glucuronide such as urine drug testing, pain prescription monitoring, toxicology analysis, or forensic testing.Specifications
67-56-1, 6801-81-6 | |
MFCD00872000 | |
100 μg/mL in methanol | |
Ampule | |
1 mL |
C21H19ClN2O8 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
462.84 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation