Learn More
CERILLIANT™ Palmitoyl-L-Carnitine (N-Methyl-D3) Solution, 500 μg/mL (90:10 Methanol:DMSO), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco A1641ML
Description
A stable-labeled internal standard of routinely tested Palmitoyl-L-Carnitine at a concentration of 500 ug/mL. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs. This stable-labeled internal standard is suitable for critical quantitative testing applications by LC/MS or GC/MS in research and clinical environments. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration value provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.Specifications
67-56-1, 202480-73-7 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
402.63 | |
-70°C | |
9.7°C (49.5°F) - closed cup |
C23H42D3NO4 | |
500 μg/mL (90:10 methanol:DMSO) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation