Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Pantothenic acid hemicalcium salt Solution, 1.0 mg/mL (90:10 Methanol:Water, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco V0621ML

Catalog No. 01-002-175

Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Pantothenic acid, also known as Vitamin B5, is an essential water-soluble B vitamin that is involved in the coenzyme A (CoA) synthesis. This certified reference solution is suitable for use as a starting material in preparation of linearity standards, calibrators, and controls for a variety of testing applications from diagnostic testing of pantothenic acid defficiency to quality control testing of pantothenic acid-based dietary supplements, fortified foods, or pharmaceutical preparations.
Specifications

Specifications

67-56-1, 137-08-6
UN 1230 - class 3 - PG 2 - Methanol, solution
238.27
1 mL
C9H16NO5 · 0.5Ca
1.0 mg/mL (90:10 methanol:water
Ampule
9.7°C (49.5°F) - closed cup
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

CERILLIANT is a registered trademark of Cerilliant Corporation