Learn More
CERILLIANT™ para-Fluorobutyryl fentanyl-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco F0740.5ML
Description
para-Fluorobutyryl fentanyl, also known as PFBF and 4-FBF, is a designer drug and fentanyl analog that is abused recreationally. This stable-labeled internal standard is suitable for quantitation of para-Fluorobutyryl fentanyl levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, forensic analysis, or isotope dilution methods.Specifications
67-56-1 | |
100 μg/mL in methanol | |
Ampule | |
9.7°C (49.5°F) - closed cup |
C1713C6H29N2OF | |
374.44 | |
0.5 mL |
Safety and Handling
H331-H370
H311
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation