Learn More
CERILLIANT™ para-Fluorofentanyl-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0700.5ML
Description
This stable-labeled internal standard is suitable for quantification of para-Fluorofentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. para-Fluorofentanyl is an opioid analgesic being an analogue of fentanyl developed by Janssen Pharmaceutica in the 1960s. para-Fluorofentanyl potently activates the human μ opioid receptor and is abused recreationally.Specifications
67-56-1 | |
100 μg/mL in methanol | |
Ampule | |
0.5 mL |
C1613C6H27N2OF | |
360.42 | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation