Learn More
CERILLIANT™ para-Fluorofentanyl-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco F0590.5ML
Description
para-Fluorofentanyl is an opioid analgesic being an analogue of fentanyl developed by Janssen Pharmaceutica in the 1960s. para-Fluorofentanyl potently activates the human μ opioid receptor and is abused recreationally. This stable-labeled internal standard is suitable for quantification of para-Fluorofentanyl levels in urine or other matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications.Specifications
67-56-1 | |
100 μg/mL in methanol | |
Ampule | |
0.5 mL |
C22H24D3FN2O | |
357.48 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation