Learn More
CERILLIANT™ para-Methylacetyl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco M2160.5ML
Description
para-Methylacetyl fentanyl is an opioid analgesic analog of fentanyl and has been sold as a designer drug on the illicit drug market. A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for para-methylacetyl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications.Specifications
67-56-1 | |
100 μg/mL in methanol (as free base) | |
Ampule | |
0.5 mL |
C22H28N2O · HCl | |
372.93 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation