Learn More
CERILLIANT™ PB-22 5-Pentanoic acid metabolite-D5 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco S0971ML
Description
A stable-labeled internal standard of PB-22 5-Pentanoic acid metabolite, suitable for use as starting material in calibrators and controls for GC/MS or LC/MS synthetic cannabinoid testing methods in forensic analysis, clinical toxicology, or urine drug testing. The 5-pentanoic acid metabolite is a primary urinary biomarker of the synthetic cannabinoids PB-22 and 5-F PB-22.Specifications
75-05-8 | |
UN 1648 3 / PGII | |
393.45 | |
-20°C | |
2°C (35.6°F) - closed cup |
C23H15D5N2O4 | |
100 μg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation