Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ PB-22 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco S0761ML
Description
PB-22, also known as QUPIC, is a synthetic cannabinoid first detected in Japanese herbal blends in 2013. The designer drug is a synthetic analog of JWH-018. This certified Snap-N-Spike™ Solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications.Specifications
75-05-8, 1400742-17-7 | |
UN 1648 3 / PGII | |
358.43 | |
-20°C | |
2°C (35.6°F) |
C23H22N2O2 | |
100 μg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
EINECSNumber : 200-835-2
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation