Learn More
CERILLIANT™ Phendimetrazine-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco P1321ML
Catalog No. 01-001-893
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
This stable-labeled internal standard is suitable for quantification of phendimetrazine levels and is applicable for use in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications by LC-MS/MS or GC/MS. Phendimetrazine, a prodrug of phenmetrazine, is a stimulant drug of the morpholine chemical class used as an appetite suppressant marketed under the names Bontril, Adipost, Anorex-SR, Appecon, Melfiat, Obezine, Phendiet, Plegine, Prelu-2, or Statobex.
Specifications
Specifications
67-56-1
UN 1230 - class 3 - PG 2 - Methanol, solution
232.76
-20°C
C12H12D5NO · HCl
100 μg/mL in methanol (as free base)
Ampule
1 mL
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation