Learn More
CERILLIANT™ Phenelzine Sulfate Solution, 1 mg/mL (0.1% 1M HCl in Methanol (v/v), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco P1391ML
Catalog No. 01-001-895
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Phenelzine, sold as Nardil or Nardelzine, is a non-selective and irreversible monoamine oxidase inhibitor (MAOI) of the hydrazine class which is used as an antidepressant and anxiolytic. This Certified Spiking Solution™ is suitable for LC/MS or GC/MS methods in phenelzine testing for forensic analysis, clinical toxicology, or urine drug testing.
Specifications
Specifications
67-56-1, 156-51-4
1 mg/mL (0.1% 1M HCl in methanol (v/v)
Ampule
1 mL
C8H12N2 · H2SO4
234.27
-70°C
9.7°C (49.5°F) - closed cup
Safety and Handling
Safety and Handling
H331-H370
H311
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation