Learn More
CERILLIANT™ Phenmetrazine-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco P1291ML
Description
Phenmetrazine, marketed under the names as Preludin, is a sympathomimetic drug that acts as a stimulant in the central nervous system primarily used as appetite suppressant. This stable-labeled internal standard is suitable for quantification of phenmetrazine levels and is applicable for use in clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research applications by LC-MS/MS or GC/MS.Specifications
67-56-1, 1246817-91-3 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
218.73 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C11H10D5NO · HCl | |
100 μg/mL in methanol (as free base) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation