Learn More
CERILLIANT™ (+/-)-Phenylephrine-d3 Hydrochloride Solution, 100 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco P0791ML
Description
An internal standard for phenylephrine, the most common over-the-counter (OTC) decongestant in the US. Over the last few years, phenylephrine has been marketed as a substitute for pseudoephedrine with tighter US government regulation of pseudoephedrine.Specifications
67-56-1, 7647-01-0 | |
UN 3286 8(6.1)(3) / PGII | |
(+\-)-Phenylephrine-D3 hydrochloride solution | |
Ampule | |
1 mL |
C9D3H11ClNO2 | |
100 μg/mL in methanol with 5% 1 M HCl (as free base) | |
206.68 | |
-20°C | |
11°C (51.8°F) - closed cup |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation