Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Pinacolyl hydrogen methylphosphonate-13C6 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco CLM66201.21.2ML
Specifications
67-56-1 | |
MFCD08275566 | |
100 μg/mL in methanol | |
Ampule | |
9.7°C (49.5°F) - closed cup |
13C6CH17O3P | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
186.14 | |
1.2 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Recommended Storage : -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation