Learn More
CERILLIANT™ Pramipexole Chiral System Suitability Solution Solution, 10 μg/mL, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco P1401ML
Specifications
110-54-3, 64-17-5 | |
10 μg/mL (85:15:0.1 Hexane:ethanol:DEA) | |
Ampule | |
1 mL |
C10H17N3S · 2HCl · H2O | |
302.26 | |
-20°C | |
-22°C (-7.6°F |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation