Learn More
CERILLIANT™ Propionyl-L-carnitine Solution, 5 mg/mL (90:10 Methanol:DMSO), Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco A1611ML
Description
A Certified Spiking Solution™ of routinely tested Propionyl-L-Carnitine at a concentration of 5 mg/mL. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs. This single-component solution certified reference material is suitable for critical quantitative applications in research and clinical environments including: for use in preparation of controls, calibrators, and linearity standards for LC-MS/MS assays; as system tuning/suitability standards; linearity verification; and internal proficiency testing. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration value provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.Specifications
67-56-1, 20064-19-1 | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
217.26 | |
-70°C | |
9.7°C (49.5°F) - closed cup |
C10H19NO4 | |
5 mg/mL (90:10 methanol:DMSO) | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation