Learn More
CERILLIANT™ (+/-)-Propoxyphene-D11 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer:  MilliporeSigma Supelco P9131ML
Catalog No. 01-001-909
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Also known as dextropropoxyphene, propoxyphene is an opioid analgesic drug intended for treatment of mild pain. In combination with either paracetamol or aspirin, dextropropoxyphene is sold under trade names including Darvocet-N™ and Darvon™. This Certified Spiking Solution™ is suitable for use as an internal standard in propoxyphene testing methods by GC/MS or LC/MS for applications in urine drug testing, pain prescription monitoring, forensic analysis, or clinical toxicology.
Specifications
Specifications
75-05-8
UN 1648 3 / PGII
(+/-)-Propoxyphene-D11 solution
Ampule
1 mL
C22D11H18NO2
100 μg/mL in acetonitrile
350.54
2 to 8°C
2°C (35.6°F) - closed cup
Safety and Handling
Safety and Handling
H332-H319
H312
EINECSNumber : 200-835-2
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
CERILLIANT is a registered trademark of Cerilliant Corporation