Learn More
CERILLIANT™ Quazepam Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco Q0061ML
Description
Quazepam, sold as Doral and Dormalin, is a long-acting benzodiazepine derivative drug for the treatment of insomnia including sleep induction and sleep maintenance. A Certified Snap-N-Spike™ Solution suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.Specifications
75-05-8, 36735-22-5 | |
UN 1648 3 / PGII | |
386.79 | |
-20°C | |
2°C (35.6°F) - closed cup |
C17H11ClF4N2S | |
1 mg/mL in acetonitrile | |
Ampule | |
1 mL |
Safety and Handling
H332-H319
H312
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation