Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ R(-)-Selegiline Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco S0031ML
Description
A Certified Spiking Solution™ suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug testing. Selegiline is an antidepressant, which belongs to the phenethylamine class of drugs. It is sold under the trade names Anipryl™, Emsam™, and Zelapar™ for treatment of Parkinson's disease, depression, and senile dementia.Specifications
67-56-1, 14611-51-9 | |
MFCD00672171 | |
1.0 mg/mL in methanol | |
187.28 | |
-20°C | |
9.7°C (49.5°F) - closed cup |
C13H17N | |
UN 1230 - class 3 - PG 2 - Methanol, solution | |
(-)-Deprenyl | |
Ampule | |
1 mL |
Safety and Handling
H331-H370
H311
EINECSNumber : 200-659-6
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation