Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Ramelteon Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco R0161ML
Description
A certified reference standard suitable as a starting material for preparation of calibrators and controls in GC/MS or LC/MS testing applications with pharmaceutical research, clinical toxicology, urine drug testing, or forensic analysis. Marketed and sold under the trade name Rozerem™, ramelteon is a newer sleep aid approved by the FDA for treatment of insomnia, particularly delayed sleep onset. The drug's mode of action is unique for a sleep drug in that it does not bind to GABA receptors like the nonbenzodiazepines zolpidem, zopiclone, and zaleplon.Specifications
75-05-8, 196597-26-9 | |
MFCD08067736 | |
1.0 mg/mL in acetonitrile | |
Ampule | |
1 mL |
C16H21NO2 | |
UN 1648 3 / PGII | |
259.34 | |
-20°C | |
2°C (35.6°F) |
Safety and Handling
H332-H319
H312
EINECSNumber : 200-835-2
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation