Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ RDX Solution, 10 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Manufacturer: MilliporeSigma Supelco ERR005S5ML
Specifications
75-05-8, 121-82-4 | |
MFCD00458725 | |
10 mg/mL in acetonitrile | |
222.12 | |
-20°C | |
2°C (35.6°F) - closed cup |
C3H6N6O6 | |
UN 3379 - DOT UN 1648 class 3 - PG 1 - Desensitized explosive, liquid, n.o.s., HI: all (not BR) | |
Hexahydro-1,3,5-trinitro-1,3,5-triazine solution | |
Ampule | |
5 mL |
Safety and Handling
H332-H319-H373
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation